Ethyl 2-amino-5-fluorobenzoate 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC535014-250MG
76601-378EA 96.82 USD
76601-378 76601-380 76601-382
Ethyl 2-amino-5-fluorobenzoate 97%
Ethyl 2-amino-5-fluorobenzoate
Formula: C₉H₁₀FNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD11042774
CAS Number: 391-93-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR