7-(Diethylamino)-4-(trifluoromethyl)coumarin

Supplier: APOLLO SCIENTIFIC
Warning

PC53487-100MG PC53487-1G PC53487-250MG
76601-292EA 32.4 USD
76601-292 76601-294 76601-296
7-(Diethylamino)-4-(trifluoromethyl)coumarin
7-(Diethylamino)-4-(trifluoromethyl)coumarin
Formula: C₁₄H₁₄F₃NO₂
Melting Pt: 78…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00057313
CAS Number: 41934-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR