FlexTech™ Y9299 Cut-Resistant Sandy Nitrile Palm Coated Gloves, Wells Lamont
Supplier: Wells Lamont
Y9299L Y9299M Y9299S Y9299XL Y9299XXL
76595-906DZ 112.38 USD
76595-906 76595-888 76595-900 76595-902 76595-890
FlexTech™ Y9299 Cut-Resistant Sandy Nitrile Palm Coated Gloves, Wells Lamont
Gloves
Protect hands with ANSI A5 cut resistance and a sandy nitrile palm.
  • 18 gauge thin glove for better dexterity
  • Touchscreen compatible
The Y9299 glove series offers ANSI Cut level A5 protection to reduce hand injuries, while providing increased abrasion resistance for extra durability and prolonged use. The ultra-lightweight liner provides a comfortable fit and excellent dexterity. The black sandy nitrile palm coating ensures a secure grip in wet, dry, or oily applications.
Certifications: ANSI level A5 cut resistance. ANSI Puncture level 4. ANSI abrasion level 4.