VWR® 2D Platform Rockers
Supplier: VWR International
76595-804 76595-814
76595-804EA 655.39 USD
76595-804 76595-814
VWR® 2D Platform Rockers
Shakers and Mixers Rotating/Rocking Shakers
VWR® 2D platform rockers are ideal for use in blotting and staining applications as well as gentle mixing of solutions and reagents.
  • Space saving design and robust construction provide durability, reliability and quiet operation
  • Large platforms with hooks for rubber strips easily accommodates many tubes, flasks and bottles
  • Models feature adjustable speed and timer
  • 2D large capacity rocker features adjustable tilt angle
  • All models suitable for use in coldrooms and standard incubator environments
  • 2 year replacement warranty
Certifications: CE Certified
Delivery information: Units supplied with dimpled rubber mat (76595-818), and rubber strips 6/pack (76595-816), instruction manual and power cord.