VWR® Chemistry-Design Diaphram Vacuum Pump
Supplier: VWR International
76595-782
76595-782EA 2770.76 USD
76595-782
VWR® Chemistry-Design Diaphram Vacuum Pump
Pumps Vacuum Diaphragm Pumps
Chemistry-design diaphram pump for distillation system.
  • Simple entry of the desired vacuum level and differential pressure
  • Precise reproducibility of set values
  • Facility to interrupt pumping program at any point
  • The chemvak system with vacuum controller automatically vents to atmosphere when switched off (this prolongs the operating life of the pump)
  • Easy venting of the system at a touch of the key pad
  • One touch resetting from controlled pumping to maximum vacuum
  • In the event of power failure the last entered values are retained
  • Continuous operation work mode
  • 50 dB noise
The vacuum pump is gas-tight, 100% oil-free, maintenance-free and quiet. All parts of the pump head in contact with gases being pumped are resistant against aggressive substances (materials: PTFE, FFKM, PVDF). New valve system is very tolerant of water vapour and condensates. Illuminated switch on pump. Over-load protection by means of thermal switch. Compact design with space saving dimensions.