TDE Series Ultra-Low Temperature Freezers with LN₂ Back-Up System, Thermo Scientific
Supplier: Thermo Fisher Scientific
TDE50086LDRLN TDE50086FARLN TDE50086FDRLN TDE50086LARLN TDE40086FDRLN TDE40086LARLN TDE40086LDRLN TDE60086FARLN TDE60086FDRLN TDE40086FARLN TDE60086LARLN TDE60086LDRLN
76595-598EA 28900 USD
76595-598 76595-572 76595-582 76595-590 76595-538 76595-544 76595-550 76595-620 76595-630 76595-530 76595-638 76595-646
TDE Series Ultra-Low Temperature Freezers with LN₂ Back-Up System, Thermo Scientific
Freezers Ultra Low Temperature Freezers
TDE series ultra-low temperature freezers are part of the Standard Performance (STP) family.
Insulation: water-blown polyurethane foam
Certifications: cULus; FDA Class II.
Delivery information: Package includes one ULT freezer, two shelves of sliding drawer racks and boxes and a factory-installed LN2 back-up system.