TDE Series Ultra-Low Freezers with Chart Recorder, Thermo Scientific
Supplier: Thermo Fisher Scientific
TDE50086LDRCR TDE50086FDRCR TDE50086LARCR TDE60086FARCR TDE60086FDRCR TDE50086FARCR TDE60086LARCR TDE60086LDRCR
76595-594EA 28400 USD
76595-594 76595-578 76595-586 76595-616 76595-626 76595-568 76595-634 76595-642
TDE Series Ultra-Low Freezers with Chart Recorder, Thermo Scientific
Freezers Ultra Low Temperature Freezers
TDE sereies ultra-low temperature freezers are part of the Standard Performance (STP) family.
Insulation: water-blown polyurethane foam
Certifications: cULus; FDA Class II.
Packaging: Pack includes one ULT freezer, two shelves of sliding drawer racks and boxes, and a factory-installed chart recorder.