TDE Series Ultra-Low Freezers with CO₂ Backup System, –40 °C, Thermo Scientific
Supplier: Thermo Fisher Scientific
TDE50040FARCO TDE40040FARCO TDE40040FDRCO TDE40040LARCO TDE40040LDRCO
76595-554EA 20200 USD
76595-554 76595-494 76595-502 76595-510 76595-518
TDE Series Ultra-Low Freezers with CO₂ Backup System, –40 °C, Thermo Scientific
Freezers Ultra Low Temperature Freezers
TDE series –40 °C ultra-low temperature freezers pack includes one ULT freezer, two shelves of sliding drawer racks and boxes and a factory-installed CO₂ back-up system.
Insulation: water-blown polyurethane foam
Certifications: EPA SNAP, Class II