Digital Hotplate, GHS-550 Group, Globe Scientific
Supplier: Globe Scientific
GHS-550H
76595-390EA 617.91 USD
76595-390
Digital Hotplate, GHS-550 Group, Globe Scientific
Heaters Hot Plates
The GHS-550 hotplate offers safety and precision for laboratory heating applications. The compact and low-profile design saves valuable benchtop space and a 7" chemical resistant glass ceramic working surface ensure years of service and easy cleanup.
  • Simple, intuitive operation
  • Small footprint saves valuable bench space
  • Powerful 1000 W heater
  • Easy-to-clean glass ceramic surface
  • Easy-to-read LED display for simple, clear control
  • Optional external temperature sensor control
Setting the heat is extremely intuitive with an LED display and control knobs, inputting to a microprocessor-controlled circuit for accurate, stable control of heating.
The hotplate is designed with a powerful 1000 W heater that provides temperatures up to 550 °C. A 'hot' warning will flash if the work plate temperature is above 50 °C. Over temperature protection is included should temperatures reach 580 °C.
Temperature accuracy: ±10 °C