PH.D.-7 Phage Display Peptide Library Kit v2, New England Biolabs
Supplier: New England Biolabs (NEB)
E8211S
76595-276EA 764 USD
76595-276
PH.D.-7 Phage Display Peptide Library Kit v2, New England Biolabs
Vectors, Plasmids and Libraries
The Ph.D.−7 phage display peptide library kit v2 is based on a combinatorial library of random heptapeptides fused to the N-terminus of a minor coat protein (pIII) of M13 phage. The peptide is followed by a short spacer (Gly-Gly-Gly) which directly precedes the wild-type pIII sequence. The library consists of ~1E09 electroporated sequences amplified once to yield approximately 100 copies of each sequence in 10 µl of the supplied phage.
  • Ready to use complex phage library (~1E09 clones)
  • Monoclonal antibody and protein G magnetic beads included for new control panning experiment
  • Panning against mouse mAb yields more reliable epitope results than former target protein
  • Does not require helper phage for amplification
  • Inherent link between phenotype and genotype allows screening of billions of clones in a single tube
The Ph.D.−7 phage display peptide library kit v2 contains the Ph.D.−7 phage display peptide library, a DYKDDDDK mouse monoclonal antibody and protein G magnetic beads for a panning control experiment, and enough −96 gIII sequencing primer for >50 sequencing reactions.