N-Boc-L-alaninol 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Boc-L-Alaninol, Boc-Alaninol, (S)-2-(BOC-amino)-1-propanol

OR61273-5G OR61273-100G OR61273-25G
76594-176EA 20.87 USD
76594-176 76594-180 76594-178
N-Boc-L-alaninol 95%
N-Boc-L-alaninol
Formula: C₈H₁₇NO₃
Melting Pt: 59…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00043121
CAS Number: 79069-13-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR