5,10,15,20-Tetrakis(2,6-dichlorophenyl)porphyrin

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5,10,15,20-Tetrakis(2,6-dichlorophenyl)porphine

OR55339-100MG
76592-424EA 172.16 USD
76592-424
5,10,15,20-Tetrakis(2,6-dichlorophenyl)porphyrin
5,10,15,20-Tetrakis(2,6-dichlorophenyl)porphyrin
Formula: C₄₄H₂₂Cl₈N₄
MW: 890.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191500
CAS Number: 37083-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR