tert-Butyl carbamate 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Carbamic acid tert-butyl ester

OR55005-25G OR55005-500G OR55005-100G
76592-176EA 20.87 USD
76592-176 76592-180 76592-178
tert-Butyl carbamate 98%
tert-Butyl carbamate
Formula: C₅H₁₁NO₂
Melting Pt: 105…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007962
CAS Number: 4248-19-5
EINECS: 224-209-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR