3-Bromo-2-nitrobenzoic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR54507-1G OR54507-250MG OR54507-5G
76591-950EA 58.04 USD
76591-950 76591-948 76591-952
3-Bromo-2-nitrobenzoic acid
3-Bromo-2-nitrobenzoic acid
Formula: C₇H₄BrNO₄
Storage Temperature: Ambient
MDL Number: MFCD10700064
CAS Number: 116529-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR