3,5-Dichloropyridine 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR5363-100G OR5363-25G
76591-716EA 45.84 USD
76591-716 76591-714
3,5-Dichloropyridine 98%
3,5-Dichloropyridine
Formula: C₅H₃Cl₂N
Melting Pt: 65…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00006376
CAS Number: 2457-47-8
EINECS: 219-537-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR