1-Decanethiol 96%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: n-Decanethiol, 1-Mercaptodecane, n-Decyl mercaptan

OR5323-100ML OR5323-2.5L OR5323-500ML
76591-652EA 33.62 USD
76591-652 76591-656 76591-654
1-Decanethiol 96%
1-Decanethiol
Formula: C₁₀H₂₂S
Boiling Pt: 114 °C/13mmHg
Melting Pt: –26 °C
Density: 0.843 g/cm³ (25 °C)
Flash Pt: 98 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004884
CAS Number: 143-10-2
EINECS: 205-584-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR