1-Decanethiol 96%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
n-Decanethiol, 1-Mercaptodecane, n-Decyl mercaptan
OR5323-100ML
OR5323-2.5L
OR5323-500ML
76591-652EA
33.62
USD
76591-652
76591-656
76591-654
1-Decanethiol 96%
1-Decanethiol
Formula:
C₁₀H₂₂S Boiling Pt: 114 °C/13mmHg Melting Pt: –26 °C Density: 0.843 g/cm³ (25 °C) Flash Pt: 98 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004884 CAS Number: 143-10-2 EINECS: 205-584-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...