Methyl 3-bromo-2-iodobenzoate 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR52724-250MG OR52724-1G
76591-336EA 75.4 USD
76591-336 76591-338
Methyl 3-bromo-2-iodobenzoate 95%
Methyl 3-bromo-2-iodobenzoate
Formula: C₈H₆BrIO₂
Storage Temperature: Ambient
MDL Number: MFCD18398801
CAS Number: 121772-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR