Medical Chemical Corporation Basic Fuchsin, Grams Counterstain, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
BF008
76591-154EA 17.44 USD
76591-154
Medical Chemical Corporation Basic Fuchsin, Grams Counterstain, Hardy Diagnostics
Stains Microbiology Stains
Basic fuchsin a stronger more intense counterstain, that produces a darker red color
Works especially well with anaerobes and other difficult to stain organisms.
Delivery information: Ship ground.