Medical Chemical Corporation Acid Alcohol, for AFB, Kinyoun or Ziehl-Neelsen, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
AA008
76591-152EA 18.46 USD
76591-152
Medical Chemical Corporation Acid Alcohol, for AFB, Kinyoun or Ziehl-Neelsen, Hardy Diagnostics
Stains Microbiology Stains
AFB decolorizer, 3% HCl in alcohol. A solution of hydrochloric acid in reagent alcohol.
Delivery information: Ship ground.