Medical Chemical Corporation Auramine-Rhodamine, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
789D8
76591-150EA 45.97 USD
76591-150
Medical Chemical Corporation Auramine-Rhodamine, Hardy Diagnostics
Stains Microbiology Stains
An aqueous solution of phenol, glycerin, isopropyl alcohol, rhodamine B and auramine O.
Delivery information: Ship ground.