Medical Chemical Corporation Acid Fast Stain Kit, Ziehl-Neelsen, for AFB Stain for Mycobacteria, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
484K
76591-148EA 57.47 USD
76591-148
Medical Chemical Corporation Acid Fast Stain Kit, Ziehl-Neelsen, for AFB Stain for Mycobacteria, Hardy Diagnostics
Stains Microbiology Stains
AFB stain kit (Ziehl Neelson) ‘Hot’ method requires heating of the slide.
Kit includes one 8 oz. bottle of each of the following: Carbol fuchsin (Ziehl Neelson), acid alcohol, methylene blue.
Delivery information: Ship ground.