Acid Fast Stain Kit with Brilliant Green Counterstain
Supplier: HARDY DIAGNOSTICS
483KB
76591-144EA 136.21 USD
76591-144
Acid Fast Stain Kit with Brilliant Green Counterstain
Stains Microbiology Stains
The acid fast stain kit is used as a differential method to detect the difference between those organisms which decolorize with treatment of acid alcohol, and those that do not, such as the mycobacteria. ‘Cold Stain’ method.
Delivery information: Ship ground.