Medical Chemical Corporation Acridine Orange Stain, 8 oz.
Supplier: HARDY DIAGNOSTICS
313B8
76591-136EA 31.23 USD
76591-136
Medical Chemical Corporation Acridine Orange Stain, 8 oz.
Stains Microbiology Stains
Acridine Orange (fluorescent stain for bacteria in body fluids) 0.025% weight by volume. An aqueous solution of acridine orange.
Delivery information: Ship ground.