3-[(Dimethylamino)methyl]-4-hydroxy-5-methoxybenzenecarbaldehyde 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-((Dimethylamino)methyl)-4-hydroxy-5-methoxybenzaldehyde

OR9894-1G
76590-894EA 20.26 USD
76590-894
3-[(Dimethylamino)methyl]-4-hydroxy-5-methoxybenzenecarbaldehyde 95%
3-[(Dimethylamino)methyl]-4-hydroxy-5-methoxybenzenecarbaldehyde
Formula: C₁₁H₁₅NO₃
Melting Pt: 140…141 °C
Storage Temperature: Ambient
MDL Number: MFCD03011572
CAS Number: 87743-10-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR