Ethyl-6-methyl-2-phenylimidazo[1,2-a]pyridine-3-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

OR9636-250MG
76590-664EA 216.74 USD
76590-664
Ethyl-6-methyl-2-phenylimidazo[1,2-a]pyridine-3-carboxylate
Ethyl 6-methyl-2-phenylimidazo[1,2-a]pyridine-3-carboxylate
Formula: C₁₇H₁₆N₂O₂
Storage Temperature: Ambient
CAS Number: 127801-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR