2,4-Dimethyl-3-hydroxypyridine 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR9029-250MG
76589-338EA 88.58 USD
76589-338 76589-336
2,4-Dimethyl-3-hydroxypyridine 97%
2,4-Dimethyl-3-hydroxypyridine
Formula: C₇H₉NO
Melting Pt: 37…40 °C
Storage Temperature: Ambient
MDL Number: MFCD08235268
CAS Number: 27296-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR