4-Aminobenzyl alcohol 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
4-(Hydroxymethyl)aniline, (4-Aminophenyl)methanol
OR9011-100G
OR9011-25G
OR9011-5G
76589-312EA
101.88
USD
76589-312
76589-310
76589-308
4-Aminobenzyl alcohol 98%
4-Aminobenzyl alcohol
Formula:
C₇H₉NO MW: 123.15 Melting Pt: 75 Storage Temperature: Refrigerator |
MDL Number:
MFCD00014782 CAS Number: 623-04-1 EINECS: 210-767-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...