4-Methoxypyridine
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR8764-25G
OR8764-5G
76589-056EA
24.46
USD
76589-056
76589-054
76589-058
4-Methoxypyridine
4-Methoxypyridine
Formula:
C₆H₇NO Boiling Pt: 191 °C Melting Pt: 4 °C Density: 1.075 g/cm³ (25 °C) Flash Pt: 74 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00674049 CAS Number: 620-08-6 EINECS: 210-624-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...