9-Bromoanthracene 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR8575-25G
OR8575-5G
76588-882EA
121.54
USD
76588-882
76588-880
76588-878
9-Bromoanthracene 98%
9-Bromoanthracene
Formula:
C₁₄H₉Br Melting Pt: 97…103 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001243 CAS Number: 1564-64-3 EINECS: 216-359-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...