2-Amino-5,6,7,8-tetrahydroquinazolin-4-ol

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Amino-4-hydroxy-5,6,7,8-tetrahydroquinazoline

OR8486-100MG
76588-794EA 22.42 USD
76588-794
2-Amino-5,6,7,8-tetrahydroquinazolin-4-ol
2-Amino-5,6,7,8-tetrahydroquinazolin-4-ol
Formula: C₈H₁₁N₃O
Storage Temperature: Ambient
MDL Number: MFCD00665850
CAS Number: 33081-07-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR