Methyl-4-bromo-3-nitrobenzoate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Bromobenzoic acid methyl ester, Methyl 4-bromo-3-nitrobenzoate

OR8177-25G OR8177-5G OR8177-1G
76588-618EA 51.96 USD
76588-618 76588-616 76588-614
Methyl-4-bromo-3-nitrobenzoate
Methyl 4-bromo-3-nitrobenzoate
Formula: C₈H₆BrNO₄
Melting Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00553089
CAS Number: 2363-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR