(S)-1,2,3,4-Tetrahydroisoquinoline-3-methanol 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
(S)-(-)-1,2,3,4-Tetrahydro-3-isoquinolinemethanol, (S)-(1,2,3,4-Tetrahydroisoquinolin-3-yl)methanol, (S)-3-Hydroxymethyl-1,2,3,4-tetrahydroisoquinoline
OR8104-1G
OR8104-25G
OR8104-5G
76588-580EA
20.32
USD
76588-580
76588-584
76588-582
(S)-1,2,3,4-Tetrahydroisoquinoline-3-methanol 97%
(S)-1,2,3,4-Tetrahydroisoquinoline-3-methanol
Formula:
C₁₀H₁₃NO Melting Pt: 114…116 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631316 CAS Number: 18881-17-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...