(S)-1,2,3,4-Tetrahydroisoquinoline-3-methanol 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (S)-(-)-1,2,3,4-Tetrahydro-3-isoquinolinemethanol, (S)-(1,2,3,4-Tetrahydroisoquinolin-3-yl)methanol, (S)-3-Hydroxymethyl-1,2,3,4-tetrahydroisoquinoline

OR8104-1G OR8104-25G OR8104-5G
76588-580EA 20.32 USD
76588-580 76588-584 76588-582
(S)-1,2,3,4-Tetrahydroisoquinoline-3-methanol 97%
(S)-1,2,3,4-Tetrahydroisoquinoline-3-methanol
Formula: C₁₀H₁₃NO
Melting Pt: 114…116 °C
Storage Temperature: Ambient
MDL Number: MFCD01631316
CAS Number: 18881-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR