3-(2,3-Dihydro-1-benzofuran-5-yl)acrylic acid 95+%

Supplier: APOLLO SCIENTIFIC
Warning

OR7839-1G
76588-320EA 54.52 USD
76588-320 76588-322
3-(2,3-Dihydro-1-benzofuran-5-yl)acrylic acid 95+%
3-(2,3-Dihydro-1-benzofuran-5-yl)acrylic acid
Formula: C₁₁H₁₀O₃
Melting Pt: 170…173 °C
Storage Temperature: Ambient
MDL Number: MFCD03002828
CAS Number: 203505-84-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR