2-[5-(Piperidin-4-yl)-1,2,4-oxadiazol-3-yl]pyrazine 95+%

Supplier: APOLLO SCIENTIFIC
Warning

OR7807-250MG
76588-284EA 87.46 USD
76588-284
2-[5-(Piperidin-4-yl)-1,2,4-oxadiazol-3-yl]pyrazine 95+%
2-[5-(Piperidin-4-yl)-1,2,4-oxadiazol-3-yl]pyrazine
Formula: C₁₁H₁₃N₅O
Melting Pt: 87…89 °C
Storage Temperature: Ambient
MDL Number: MFCD03426077
CAS Number: 849925-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR