4-Carboxyphenylboronic acid
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
4-Boronobenzoic Acid, Ark Pharm, 4-Carboxybenzeneboronic acid
OR7732-100G
OR7732-25G
OR7732-5G
76588-184EA
88.62
USD
76588-184
76588-182
76588-180
4-Carboxyphenylboronic acid
4-Carboxyphenylboronic acid
Formula:
C₇H₇BO₄ Melting Pt: 220 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00151801 CAS Number: 14047-29-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...