[1,2,4]Triazol-1-ylacetic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR7103-1G OR7103-5G
76587-670EA 100.84 USD
76587-670 76587-672
[1,2,4]Triazol-1-ylacetic acid 98%
[1,2,4]Triazol-1-ylacetic acid
Formula: C₄H₅N₃O₂
Melting Pt: 201…205 °C
Storage Temperature: Ambient
MDL Number: MFCD00036051
CAS Number: 28711-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR