3-Bromo-7-nitro-1H-indazole
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR6862T-1G
OR6862T-5G
76587-182EA
20.26
USD
76587-182
76587-180
76587-184
3-Bromo-7-nitro-1H-indazole
3-Bromo-7-nitro-1H-indazole
A more potent inhibitor of rat cerebellar nitric oxide synthase than 7-nitroindazole.Harmful to reproduction.
Formula:
C₇H₄BrN₃O₂ Melting Pt: 177…178 °C Storage Temperature: Ambient |
MDL Number:
MFCD00159910 CAS Number: 74209-34-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...