2-Bromo-5-chloropyridine 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR5182-100G OR5182-10G OR5182-25G
76586-794EA 57.04 USD
76586-794 76586-790 76586-792
2-Bromo-5-chloropyridine 98%
2-Bromo-5-chloropyridine
Formula: C₅H₃BrClN
Melting Pt: 65…69 °C
Storage Temperature: Ambient
MDL Number: MFCD00234006
CAS Number: 40473-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR