Gallic acid, anhydrous 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3,4,5-Trihydroxybenzoic acid

OR51754-1KG OR51754-250G OR51754-5KG
76586-692EA 89.64 USD
76586-692 76586-690 76586-694
Gallic acid, anhydrous 99%
Gallic acid
Formula: C₇H₆O₅
MW: 170.12 g/mol
Boiling Pt: 501
Melting Pt: 251 °C (dec.)
Density: 1.694 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002510
CAS Number: 149-91-7
EINECS: 205-749-9
Merck Index: 13,04366

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR