2-Chloronicotinic acid 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
2-Chloro-3-pyridinecarboxylic acid, 2-Chloropyridine-3-carboxylic acid
OR5074-100G
OR5074-1KG
OR5074-500G
76586-270EA
20.87
USD
76586-270
76586-274
76586-272
2-Chloronicotinic acid 98%
2-Chloronicotinic acid
Formula:
C₆H₄ClNO₂ Melting Pt: 176…178 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00006236 CAS Number: 2942-59-8 EINECS: 220-937-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...