2-Chloronicotinic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Chloro-3-pyridinecarboxylic acid, 2-Chloropyridine-3-carboxylic acid

OR5074-100G OR5074-1KG OR5074-500G
76586-270EA 20.87 USD
76586-270 76586-274 76586-272
2-Chloronicotinic acid 98%
2-Chloronicotinic acid
Formula: C₆H₄ClNO₂
Melting Pt: 176…178 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006236
CAS Number: 2942-59-8
EINECS: 220-937-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR