2-Bromo-4-hydroxy-5-methoxyphenylacetic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR4895-100MG OR4895-500MG
76585-864EA 195.76 USD
76585-864 76585-866
2-Bromo-4-hydroxy-5-methoxyphenylacetic acid
2-Bromo-4-hydroxy-5-methoxyphenylacetic acid
Formula: C₉H₉BrO₄
Melting Pt: 174…177 °C
Storage Temperature: Ambient
MDL Number: MFCD00079715
CAS Number: 181517-98-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR