3-Nitro-1H-1,2,4-triazole

Supplier: APOLLO SCIENTIFIC
Warning

OR4862-25G OR4862-5G
76585-856EA 30.56 USD
76585-856 76585-854
3-Nitro-1H-1,2,4-triazole
3-Nitro-1H-1,2,4-triazole
Formula: C₂H₂N₄O₂
Melting Pt: 210 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00009749
CAS Number: 24807-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR