7-Ethoxy-4-methylcoumarin 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR4853-100G OR4853-25G OR4853-5G
76585-850EA 110.02 USD
76585-850 76585-848 76585-846
7-Ethoxy-4-methylcoumarin 98%
7-Ethoxy-4-methylcoumarin
Formula: C₁₂H₁₂O₃
Melting Pt: 113…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00016971
CAS Number: 87-05-8
EINECS: 201-721-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR