2-Bromo-6-chlorophenylacetylene 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR48117-1G OR48117-250MG
76585-748EA 80.48 USD
76585-748 76585-746
2-Bromo-6-chlorophenylacetylene 97%
2-Bromo-6-chlorophenylacetylene
Formula: C₈H₄BrCl
Storage Temperature: Ambient
MDL Number: MFCD31700579
CAS Number: 1856321-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR