5-Amino-4-bromo-2-chlorophenol 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR48111-1G OR48111-250MG
76585-740EA 543.96 USD
76585-740 76585-738
5-Amino-4-bromo-2-chlorophenol 95%
5-Amino-4-bromo-2-chlorophenol
Formula: C₆H₅BrClNO
Storage Temperature: Ambient
MDL Number: MFCD27942409
CAS Number: 1232505-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR