4-(Isobutyro)thioanisole

Supplier: APOLLO SCIENTIFIC
Warning

OR4659-100G OR4659-25G
76585-136EA 260.14 USD
76585-136 76585-134
4-(Isobutyro)thioanisole
4-(Isobutyro)thioanisole
Formula: C₁₁H₁₄OS
Boiling Pt: 122…124 °C/2mmHg
Melting Pt: 32…33 °C
Storage Temperature: Ambient
MDL Number: MFCD03094661
CAS Number: 53207-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR