3-Methoxypyridin-4-amine

Supplier: APOLLO SCIENTIFIC
Warning

OR46502-100MG OR46502-250MG OR46502-500MG
76585-036EA 63.16 USD
76585-036 76585-038 76585-040
3-Methoxypyridin-4-amine
3-Methoxypyridin-4-amine
Formula: C6H8N2O
Storage Temperature: Ambient
MDL Number: MFCD09991938
CAS Number: 52334-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR